CAS RN 57234-29-4
IUPAC Name:
1,3-bis(methoxymethoxy)benzene
Synonyms:
1,3-bis(methoxymethoxy)benzene
Molecular Weight (g/mol):
198.22
Molecular Formula:
C10H14O4
InChi Key:
TZZPMRGYEPMHLA-UHFFFAOYSA-N
SMILES:
COCOC1=CC(OCOC)=CC=C1
1
–
1
of
1
results
1
1,3-Bis(methoxymethoxy)benzene 98.0+%, TCI America™
CAS: 57234-29-4 Molecular Formula: C10H14O4 Molecular Weight (g/mol): 198.22 MDL Number: MFCD27925713 InChI Key: TZZPMRGYEPMHLA-UHFFFAOYSA-N Synonym: Resorcinol Bis(methoxymethyl) Ether PubChem CID: 12222682 IUPAC Name: 1,3-bis(methoxymethoxy)benzene SMILES: COCOC1=CC(OCOC)=CC=C1