accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNNH

CAS RN 57260-67-0

IUPAC Name: 1-(3,4-dichlorophenyl)piperazine
Synonyms: 1-(3,4-dichlorophenyl)piperazine
Molecular Weight (g/mol): 231.12
Molecular Formula: C10H12Cl2N2
InChi Key: PXFJLKKZSWWVRX-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)N1CCNCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results