Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,4-Dichlorophenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. D40215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40215G Supplier TCI America Supplier No. D40215G

missing translation for 'validMainframeMsg'

Specifications

CAS 57260-67-0
Melting Point 66°C
Color White-Yellow
Boiling Point 202°C
Molecular Formula C10H12Cl2N2
Quantity 5 g
MDL Number MFCD00023128
UN Number 3259
Synonym 1-3,4-dichlorophenyl piperazine, n-3,4-dichlorophenyl piperazine, 1-3,4-dichlorophenyl-piperazine, piperazine, 1-3,4-dichlorophenyl, 3,4-dichlorophenyl piperazine, 1-3,4-dichloro-phenyl-piperazine, 1-3,4dichlorophenyl piperazine, 1-3,4-dichlorphenyl piperazin, pubchem8584, acmc-1b0ni
InChI Key PXFJLKKZSWWVRX-UHFFFAOYSA-N
SMILES ClC1=CC=C(C=C1Cl)N1CCNCC1
IUPAC Name 1-(3,4-dichlorophenyl)piperazine
Molecular Weight (g/mol) 231.12
PubChem CID 93601
Formula Weight 231.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3,4-Dichlorophenyl)piperazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.