accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH3COCH3

CAS RN 573-11-5

IUPAC Name: 2,3,4-trimethoxybenzoate
Synonyms: 2,3,4-trimethoxybenzoate
Molecular Weight (g/mol): 211.19
Molecular Formula: C10H11O5
InChi Key: HZNQSWJZTWOTKM-UHFFFAOYSA-M
SMILES: COC1=CC=C(C([O-])=O)C(OC)=C1OC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)