accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONFFFNOO

CAS RN 573759-00-9

IUPAC Name: 4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
Synonyms: 4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
Molecular Weight (g/mol): 232.12
Molecular Formula: C8H3F3N2O3
InChi Key: MVYGNYLHXPFQJV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2N=C(OC2=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results