accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOS(Z)

CAS RN 57409-51-5

IUPAC Name: methyl (2Z)-2-hydroxy-4-oxo-4-(thiophen-2-yl)but-2-enoate
Synonyms: methyl (2Z)-2-hydroxy-4-oxo-4-(thiophen-2-yl)but-2-enoate
Molecular Weight (g/mol): 212.22
Molecular Formula: C9H8O4S
InChi Key: TZNHHDJRXUBHRB-ALCCZGGFSA-N
SMILES: COC(=O)C(\O)=C\C(=O)C1=CC=CS1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)