Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-Dioxo-4-(2-thienyl)butyrate 98.0+%, TCI America™
SDP

Catalog No. M29361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M29361G Supplier TCI America Supplier No. M29361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57409-51-5
Molecular Formula C9H8O4S
Molecular Weight (g/mol) 212.22
MDL Number MFCD01249731
InChI Key TZNHHDJRXUBHRB-ALCCZGGFSA-N
Synonym methyl 2,4-dioxo-4-thiophen-2-yl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid methyl ester, methyl 2,4-dioxo-4-thien-2-ylbutanoate, methyl 2,4-dioxo-4-2-thienyl butanoate, methyl 2,4-dioxo-4-2-thienyl butyrate, methyl 2-thienoylpyruvate, methyl 4-2-thienyl-2,4-dioxobutanoate, methyl 2,4-dioxo-4-thien-2-yl butanoate, methyl2,4-dioxo-4-thiophen-2-yl butanoate, 2-thiophenebutanoicacid, a,g-dioxo-, methyl ester
PubChem CID 2771673
IUPAC Name methyl (2Z)-2-hydroxy-4-oxo-4-(thiophen-2-yl)but-2-enoate
SMILES COC(=O)C(\O)=C\C(=O)C1=CC=CS1

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,4-Dioxo-4-(2-thienyl)butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.