accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57414-85-4

IUPAC Name: ethyl 3-amino-2-methylbenzoate
Synonyms: ethyl 3-amino-2-methylbenzoate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: RZDRJEHTKWQFQO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C(N)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results