Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Amino-2-methylbenzoate 98.0+%, TCI America™
SDP

Catalog No. A15745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15745G 5 g
A157425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15745G Supplier TCI America Supplier No. A15745G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57414-85-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00957246
InChI Key RZDRJEHTKWQFQO-UHFFFAOYSA-N
Synonym 3-Amino-2-methylbenzoic Acid Ethyl Ester, Ethyl 3-Amino-o-toluate, 3-Amino-o-toluic Acid Ethyl Ester
PubChem CID 4313471
IUPAC Name ethyl 3-amino-2-methylbenzoate
SMILES CCOC(=O)C1=C(C(=CC=C1)N)C

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Amino-2-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.