accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3NOOOClCl

CAS RN 5742-17-6

IUPAC Name: propan-2-aminium 2-(2,4-dichlorophenoxy)acetate
Synonyms: isopropylaminium 2,4-dichlorophenoxyacetate
Molecular Weight (g/mol): 280.15
Molecular Formula: C11H15Cl2NO3
InChi Key: HXMBBZAHRWCZOX-UHFFFAOYSA-N
SMILES: CC(C)[NH3+].[O-]C(=O)COC1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results