accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5743-26-0

IUPAC Name: calcium diacetate hydrate
Synonyms: calcium acetate monohydrate Ca(OAc)2.H2O calcium acetate hydrate (1:2:1) calcium diacetate monohydrate
Molecular Weight (g/mol): 176.18
Molecular Formula: C4H8CaO5
InChi Key: XQKKWWCELHKGKB-UHFFFAOYSA-L
SMILES: O.[Ca++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (11)
  • (2)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (8)
Grade 
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (5)
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)
  • (4)
  • (3)
  • (5)
  • (1)
  • (11)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (8)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (5)