Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Acetate, Monohydrate
SCI_ED

Catalog No. S25218A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25218A 500 g Lab Grade
S25218 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25218A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.181
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
PubChem CID 82163
ChEBI CHEBI:59199
IUPAC Name calcium;diacetate;hydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

Specifications

Color White
Quantity 500 g
Assay Percent Range 1
Linear Formula Ca(C2H3O2)2·H2O
Formula Weight 176.13
Grade Laboratory
Packaging Lab Grade
Density 1.5g/mL
Chemical Name or Material Calcium Acetate, Monohydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.