accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OCaH3COOOH3COOO

CAS RN 5743-49-7

IUPAC Name: calcium bis(4-oxopentanoate) dihydrate
Synonyms: calcium dihydrate dilevulinate
Molecular Weight (g/mol): 306.32
Molecular Formula: C10H18CaO8
InChi Key: WQFDRFDIWAMOPA-UHFFFAOYSA-L
SMILES: O.O.[Ca++].CC(=O)CCC([O-])=O.CC(=O)CCC([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results