Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Levulinate Dihydrate, TCI America™
SDP

Catalog No. L004325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L004325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L004325G Supplier TCI America Supplier No. L004325G

Chemical Identifiers

CAS 5743-49-7
Molecular Formula C10H20CaO8
Molecular Weight (g/mol) 308.34
MDL Number MFCD00045959
InChI Key GYALXOMMCMBYJS-UHFFFAOYSA-N
Synonym 3-Acetylpropionic Acid Calcium Salt, Levulinic Acid Calcium Salt, 4-Oxovaleric Acid Calcium Salt
PubChem CID 129894278
IUPAC Name calcium;4-oxopentanoic acid;dihydrate
SMILES CC(=O)CCC(=O)O.CC(=O)CCC(=O)O.O.O.[Ca]

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 25 g
Formula Weight 270.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Levulinate Dihydrate
RTECSNumber SA4000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.