accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3NH3CN

CAS RN 5744-51-4

IUPAC Name: ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate
Synonyms: ethyl 1,5-dimethylpyrazole-3-carboxylate
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: OJPXVXXMBWKEAT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NN(C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results