accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOF

CAS RN 574715-85-8

IUPAC Name: N,N-dicyclohexyl-2-fluorobenzamide
Synonyms: N,N-dicyclohexyl-2-fluorobenzamide
Molecular Weight (g/mol): 303.42
Molecular Formula: C19H26FNO
InChi Key: ZHVPNXNRPGNFGS-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(=O)N(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results