accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSSO

CAS RN 57500-00-2

IUPAC Name: 2-[(methyldisulfanyl)methyl]furan
Synonyms: 2-[(methyldisulfanyl)methyl]furan
Molecular Weight (g/mol): 160.25
Molecular Formula: C6H8OS2
InChi Key: CLSLQQCDHOZMDT-UHFFFAOYSA-N
SMILES: CSSCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results