Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl Methyl Disulfide 98.0+%, TCI America™
SDP

Catalog No. F05615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05615G Supplier TCI America Supplier No. F05615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 57500-00-2
Color Yellow
Boiling Point 61°C
Molecular Formula C6H8OS2
Quantity 5 g
MDL Number MFCD00012326
InChI Key CLSLQQCDHOZMDT-UHFFFAOYSA-N
SMILES CSSCC1=CC=CO1
IUPAC Name 2-[(methyldisulfanyl)methyl]furan
Molecular Weight (g/mol) 160.249
PubChem CID 62131
Formula Weight 160.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Methyl Disulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.