accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNHN

CAS RN 57500-07-9

IUPAC Name: 6-(benzyloxy)-7H-purine
Synonyms: 6-(benzyloxy)-7H-purine
Molecular Weight (g/mol): 226.24
Molecular Formula: C12H10N4O
InChi Key: ZZZXGPGVDJDFCJ-UHFFFAOYSA-N
SMILES: C(OC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results