accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOOCH3H3CCH3OHO

CAS RN 57521-85-4

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: INWOAUUPYIXDHN-UHFFFAOYNA-N
SMILES: CCCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results