Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-norvaline dicyclohexylammonium salt, 98%, Thermo Scientific™

Catalog No. AAH6179603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6179603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6179603 Supplier Thermo Scientific Chemicals Supplier No. H6179603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57521-85-4
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD11041002
InChI Key INWOAUUPYIXDHN-SSDOTTSWSA-N
Synonym boc-d-norvaline, boc-d-nva-oh, r-2-tert-butoxycarbonyl amino pentanoic acid, 2r-2-tert-butoxycarbonyl amino pentanoic acid, boc-d-2-aminovaleric acid, n-t-butoxycarbonyl-d-norvaline, n-tert-butoxycarbonyl-d-norvaline, boc-d-nva-oh tlc, r-2-tert-butoxycarbonylamino-pentanoic acid, d-norvaline, n-1,1-dimethylethoxy carbonyl
PubChem CID 6995000
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CCCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Color White
Quantity 1 g
Formula Weight 398.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-D-norvaline dicyclohexylammonium salt
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.