accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrH3C

CAS RN 576-23-8

IUPAC Name: 1-bromo-2,3-dimethylbenzene
Synonyms: 3-bromo-1,2-dimethylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: WLPXNBYWDDYJTN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Br)=C1C
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)