Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-o-xylene 98.0+%, TCI America™
SDP

Catalog No. B12825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12825G 5 g
B128225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12825G Supplier TCI America Supplier No. B12825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 576-23-8 | C8H9Br | 185.06 g/mol

Chemical Identifiers

CAS 576-23-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000069
InChI Key WLPXNBYWDDYJTN-UHFFFAOYSA-N
Synonym 3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
PubChem CID 68472
IUPAC Name 1-bromo-2,3-dimethylbenzene
SMILES CC1=CC=CC(Br)=C1C

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 214°C
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 3-Bromo-o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.