accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 5764-85-2

IUPAC Name: ethyl 3-hydroxy-3-phenylpropanoate
Synonyms: ethyl 3-hydroxy-3-phenylpropanoate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: DVIBDQWVFHDBOP-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results