accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClH3CH2N

CAS RN 5769-32-4

IUPAC Name: 4-chloro-2,6-dimethylaniline
Synonyms: 4-chloro-2,6-dimethylaniline
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: JCLZLZKUISPXDC-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(C)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results