accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OO

CAS RN 57699-45-3

IUPAC Name: 4-(tert-butoxy)benzaldehyde
Synonyms: 4-(tert-butoxy)benzaldehyde
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: VWSFZYXXQDKXKQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results