accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 57718-18-0

IUPAC Name: (non-1-yn-1-yl)benzene
Synonyms: non-1-yn-1-ylbenzene
Molecular Weight (g/mol): 200.33
Molecular Formula: C15H20
InChi Key: ZRQWNFWWPPYNFA-UHFFFAOYSA-N
SMILES: CCCCCCCC#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results