Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-nonyne 98.0+%, TCI America™
SDP

Catalog No. P03635ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03635ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P03635ML Supplier TCI America Supplier No. P03635ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57718-18-0
Molecular Formula C15H20
Molecular Weight (g/mol) 200.33
MDL Number MFCD00048944
InChI Key ZRQWNFWWPPYNFA-UHFFFAOYSA-N
Synonym 1-Heptyl-2-phenylacetylene
PubChem CID 143501
IUPAC Name (non-1-yn-1-yl)benzene
SMILES CCCCCCCC#CC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 200.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-nonyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.