accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5779-72-6

IUPAC Name: 2,4,5-trimethylbenzaldehyde
Synonyms: 2,4,5-trimethylbenzaldehyde
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: LROJZZICACKNJL-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=O)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results