Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trimethylbenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. T141125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T141125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T141125G Supplier TCI America Supplier No. T141125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5779-72-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00017713
InChI Key LROJZZICACKNJL-UHFFFAOYSA-N
PubChem CID 22013
IUPAC Name 2,4,5-trimethylbenzaldehyde
SMILES CC1=CC(C)=C(C=O)C=C1C

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 243°C
Quantity 25 g
Formula Weight 148.21
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,4,5-Trimethylbenzaldehyde
EINECSNumber (3)-3816
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.