accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3

CAS RN 57802-79-6

IUPAC Name: (4-butylphenyl)methanaminium
Synonyms: (4-butylphenyl)methanaminium
Molecular Weight (g/mol): 164.27
Molecular Formula: C11H18N
InChi Key: IBVGSPOHLFKLHM-UHFFFAOYSA-O
SMILES: CCCCC1=CC=C(C[NH3+])C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results