accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57803-73-3

IUPAC Name: 5-methylheptan-1-ol
Synonyms: 5-methylheptan-1-ol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: KFARNLMRENFOHE-UHFFFAOYNA-N
SMILES: CCC(C)CCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results