Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-5-Methyl-1-heptanol 97.0+%, TCI America™
SDP

Catalog No. M09651ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09651ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09651ML Supplier TCI America Supplier No. M09651ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57803-73-3
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00135168
InChI Key KFARNLMRENFOHE-QMMMGPOBSA-N
PubChem CID 13218084
IUPAC Name (5S)-5-methylheptan-1-ol
SMILES CCC(C)CCCCO

Specifications

Color Yellow
Boiling Point 84°C
Quantity 1 mL
Formula Weight 130.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-5-Methyl-1-heptanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.