accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFSS

CAS RN 57842-27-0

IUPAC Name: 1λ⁴,3-benzodithiol-1-ylium; tetrafluoroboranuide
Synonyms: 1λ⁴,3-benzodithiol-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 240.04
Molecular Formula: C7H5BF4S2
InChi Key: CUSWPJQKCZMDPY-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.S1C=[S+]C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results