Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzodithiolylium Tetrafluoroborate 96.0+%, TCI America™
SDP

Catalog No. B11515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11515G Supplier TCI America Supplier No. B11515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57842-27-0
Molecular Formula C7H5BF4S2
Molecular Weight (g/mol) 240.041
MDL Number MFCD00070613
InChI Key CUSWPJQKCZMDPY-UHFFFAOYSA-N
Synonym 1,3-benzodithiolylium tetrafluoroborate, benzo d 1,3 dithiol-1-ium tetrafluoroborate, 1,3-benzodithiolium tetrafluoroborate, 1,3-benzodithiol-3-ium tetrafluoroborate, 1,3-benzodithiol-2-ylium tetrafluoroborate, 1??,3-benzodithiol-1-ylium tetrafluoroborate, 2h-1,3-benzodithiol-2-ylium tetrafluoroborate, c7h5s2.bf4, 1,3-benzolithiolylium tetrafluoroborate
PubChem CID 2735971
IUPAC Name 1,3-benzodithiol-3-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1=CC=C2C(=C1)SC=[S+]2

Specifications

Quantity 5 g
UN Number 1759
Formula Weight 240.04
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Benzodithiolylium Tetrafluoroborate [Hydroxyl-Protecting Agent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.