accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57876-69-4

IUPAC Name: 2-chloro-3-methylquinoline
Synonyms: 2-chloro-3-methylquinoline
Molecular Weight (g/mol): 177.63
Molecular Formula: C10H8ClN
InChi Key: UMYPPRQNLXTIEQ-UHFFFAOYSA-N
SMILES: CC1=C(Cl)N=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results