Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-3-methylquinoline, 97%, Thermo Scientific™

Catalog No. NRB00907DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
NRB00907DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NRB00907DA Supplier Thermo Scientific Chemicals Supplier No. NRB00907DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57876-69-4
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD00100265
InChI Key UMYPPRQNLXTIEQ-UHFFFAOYSA-N
Synonym 2-chloro-3-methyl-quinoline, quinoline, 2-chloro-3-methyl, zlchem 105, pubchem12431, acmc-1ayrn, methyl 2-chloroquinoline, 2-chloro-3-methylquinolin, 3-methyl-2-chloroquinoline, 2-cloro-3-methyl-quinoline, 2-chloro-3-methyl quinoline
PubChem CID 339082
IUPAC Name 2-chloro-3-methylquinoline
SMILES CC1=CC2=CC=CC=C2N=C1Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 2-chloro-3-methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.