accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5788-17-0

IUPAC Name: methyl (2E)-3-methoxyprop-2-enoate
Synonyms: methyl (2E)-3-methoxyprop-2-enoate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: AUTCCPQKLPMHDN-ONEGZZNKSA-N
SMILES: CO\C=C\C(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)