Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-3-methoxyacrylate 95%, Thermo Scientific™

Catalog No. AC329711000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC329711000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC329711000 Supplier Thermo Scientific Chemicals Supplier No. 329711000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5788-17-0
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
InChI Key AUTCCPQKLPMHDN-ONEGZZNKSA-N
Synonym methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
PubChem CID 5323651
IUPAC Name methyl (E)-3-methoxyprop-2-enoate
SMILES COC=CC(=O)OC

Specifications

Color Colorless
Density 1.08
Infrared Spectrum Authentic
Refractive Index 1.4470 to 1.4510 (20°C, 589nm)
Linear Formula CH3OCH=CHCO2CH3
Quantity 100 mL
Formula Weight 116.12
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Methyl trans-3-methoxyacrylate, 95%
Hazard Category LACHRYMATOR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.