CAS RN 5789-30-0
IUPAC Name:
(1,2-dibromo-2-phenylethyl)benzene
Synonyms:
(1,2-dibromo-2-phenylethyl)benzene
Molecular Weight (g/mol):
340.06
Molecular Formula:
C14H12Br2
InChi Key:
GKESIQQTGWVOLH-UHFFFAOYNA-N
SMILES:
BrC(C(Br)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1,2-Dibromo-1,2-diphenylethane, 96%, Thermo Scientific Chemicals
1,2-Dibromo-1,2-diphenylethane, 96%, Thermo Scientific Chemicals
CAS: 5789-30-0 Molecular Formula: C14H12Br2 Molecular Weight (g/mol): 340.05 MDL Number: MFCD00000137 InChI Key: GKESIQQTGWVOLH-UHFFFAOYSA-N Synonym: 1,2-dibromo-1,2-diphenylethane,stilbene dibromide,1,2-dibromo-2-phenylethyl benzene,benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis,meso-dibromostilbene,benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis-, r*,s*,alpha,alpha-dibromobibenzyl,bibenzyl,,a,,a'-dibromo,bibenzyl, alpha,alpha'-dibromo,acmc-1ao3z PubChem CID: 93010 IUPAC Name: (1,2-dibromo-2-phenylethyl)benzene SMILES: C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br