Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-1,2-diphenylethane, 96%, Thermo Scientific Chemicals

Catalog No. AC147950250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147950250 25 g Glass bottle
AC147950050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC147950250 Supplier Thermo Scientific Chemicals Supplier No. 147950250
  • $374.00
    $314.65 / Each of 1

    Save $59.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5789-30-0
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.05
MDL Number MFCD00000137
InChI Key GKESIQQTGWVOLH-UHFFFAOYSA-N
Synonym 1,2-dibromo-1,2-diphenylethane, stilbene dibromide, 1,2-dibromo-2-phenylethyl benzene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis, meso-dibromostilbene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis-, r*,s*, alpha,alpha-dibromobibenzyl, bibenzyl,, a,, a'-dibromo, bibenzyl, alpha,alpha'-dibromo, acmc-1ao3z
PubChem CID 93010
IUPAC Name (1,2-dibromo-2-phenylethyl)benzene
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 235°C to 241°C
Color Orange-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CH(Br)C6H5
Quantity 25 g
Beilstein 05, 602
Formula Weight 340.05
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1, 2-Dibromo-1, 2-diphenylethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 227-324-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.