accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3O

CAS RN 579-10-2

IUPAC Name: N-methyl-N-phenylacetamide
Synonyms: N-methylacetanilide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: LMTGCJANOQOGPI-UHFFFAOYSA-N
SMILES: CN(C(C)=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results