Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylacetanilide 98.0+%, TCI America™
SDP

Catalog No. M013425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M013425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M013425G Supplier TCI America Supplier No. M013425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 579-10-2
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00026181
InChI Key LMTGCJANOQOGPI-UHFFFAOYSA-N
Synonym N-Acetylmethylaniline
PubChem CID 11364
IUPAC Name N-methyl-N-phenylacetamide
SMILES CN(C(C)=O)C1=CC=CC=C1

Specifications

Melting Point 101°C
Color White-Yellow
Boiling Point 146°C
Quantity 25 g
UN Number 2811
Formula Weight 149.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylacetanilide
EINECSNumber (3)-0113
RTECSNumber AE4900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.