accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaOHOHOHOOOHOO

CAS RN 5793-88-4

IUPAC Name: calcium 2,3,4,5-tetrahydroxyhexanedioate
Synonyms: calcium D-saccharate
Molecular Weight (g/mol): 248.2
Molecular Formula: C6H8CaO8
InChi Key: UGZVNIRNPPEDHM-UHFFFAOYNA-L
SMILES: [Ca++].OC(C(O)C(O)C([O-])=O)C(O)C([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results