accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaOOS

CAS RN 5793-98-6

IUPAC Name: calcium 2-sulfanidylacetate
Synonyms: calcium thioglycolate(2-)
Molecular Weight (g/mol): 130.17
Molecular Formula: C2H2CaO2S
InChi Key: QVCFQEFEMBCIOI-UHFFFAOYSA-L
SMILES: [Ca++].[O-]C(=O)C[S-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results