accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHFNH2

CAS RN 579474-47-8

IUPAC Name: tert-butyl N-(2-amino-4-fluorophenyl)carbamate
Synonyms: tert-butyl N-(2-amino-4-fluorophenyl)carbamate
Molecular Weight (g/mol): 226.25
Molecular Formula: C11H15FN2O2
InChi Key: IBKSOWUYKOORFF-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=C(N)C=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results