Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. B4236200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4236200MG 200 mg
B42361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4236200MG Supplier TCI America Supplier No. B4236200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 579474-47-8
Molecular Formula C11H15FN2O2
Molecular Weight (g/mol) 226.25
MDL Number MFCD09701246
InChI Key IBKSOWUYKOORFF-UHFFFAOYSA-N
Synonym N1-Boc-4-fluoro-1,2-phenylenediamine, tert-Butyl (2-Amino-4-fluorophenyl)carbamate, (2-Amino-4-fluorophenyl)carbamic Acid tert-Butyl Ester
PubChem CID 24688763
IUPAC Name tert-butyl N-(2-amino-4-fluorophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=C(N)C=C(F)C=C1

Specifications

Melting Point 137°C
Quantity 200 mg
Formula Weight 226.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N1-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.