accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHHOHOO

CAS RN 5796-17-8

IUPAC Name: 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Synonyms: 3',4'-dihydroxyphenylalanine 3,4-dihydroxyphenyl-L-alanine helfo dopa 3-hydroxytyrosine dopicar biodopa cerepap DL-dihydroxyphenylalanine L-(o-dihydroxyphenyl)alanine sobiodopa
Molecular Weight (g/mol): 197.19
Molecular Formula: C9H11NO4
InChi Key: WTDRDQBEARUVNC-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(O)C(O)=C1)C(O)=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)