accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFH3CSSCH3H3C

CAS RN 5799-67-7

IUPAC Name: tetrafluoroboranuide; trimethyldisulfanium
Synonyms: trimethyldisulfanium tetrafluoroborate
Molecular Weight (g/mol): 196.03
Molecular Formula: C3H9BF4S2
InChi Key: LUOOXKXNWLQGLY-UHFFFAOYSA-N
SMILES: CS[S+](C)C.F[B-](F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)