Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl(methylthio)sulfonium Tetrafluoroborate 97.0+%, TCI America™
SDP

Catalog No. D19455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19455G 5 g
D19451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19455G Supplier TCI America Supplier No. D19455G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5799-67-7
Molecular Formula C3H9BF4S2
Molecular Weight (g/mol) 196.03
MDL Number MFCD00011806
InChI Key LUOOXKXNWLQGLY-UHFFFAOYSA-N
Synonym DMTSF
PubChem CID 2737114
IUPAC Name tetrafluoroboranuide; trimethyldisulfanium
SMILES CS[S+](C)C.F[B-](F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 196.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl(methylthio)sulfonium Tetrafluoroborate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.